Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUBCNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102041.2 | NP_001095511.1 | 4 - 5 | 971 | 58 | |
GenBank mRNA | BC151662.1 | - | 4 - 5 | 972 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |