Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102053.2 | NP_001095523.2 | 2 - 3 | 2064 | 86 | |
GenBank mRNA | BC149084.1 | - | 4 - 5 | 577 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |