Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105345.2 | NP_001098815.1 | 1 - 2 | 212 | 83 | |
GenBank mRNA | BC151519.1 | - | 1 - 2 | 213 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |