Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098127.1 | NP_001091596.1 | 5 - 6 | 568 | 56 | |
GenBank mRNA | BC140521.1 | - | 5 - 6 | 568 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |