Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099039.2 | NP_001092509.1 | 2 - 3 | 330 | 68 | |
GenBank mRNA | BC142315.1 | - | 2 - 3 | 332 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |