Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099161.2 | NP_001092631.1 | 1 - 2 | 487 | 95 | |
GenBank mRNA | BC142451.1 | - | 1 - 2 | 488 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |