Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVI2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099166.2 | NP_001092636.1 | 1 - 2 | 138 | 123 | |
GenBank mRNA | BC142467.1 | - | 1 - 2 | 139 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |