Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101125.1 | NP_001094595.1 | 7 - 8 | 894 | 73 | |
GenBank mRNA | BC150078.1 | - | 7 - 8 | 894 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |