Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101148.2 | NP_001094618.1 | 1 - 2 | 88 | 123 | |
GenBank mRNA | BC149358.1 | - | 1 - 2 | 89 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |