Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102150.1 | NP_001095620.1 | 4 - 5 | 484 | 80 | |
GenBank mRNA | BC150088.1 | - | 4 - 5 | 484 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |