Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102373.1 | NP_001095843.1 | 1 - 2 | 480 | 119 | |
GenBank mRNA | BC114690.1 | - | 1 - 2 | 480 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |