Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103291.1 | NP_001096761.1 | 13 - 14 | 1941 | 150 | |
GenBank mRNA | BC123438.1 | - | 13 - 14 | 1941 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |