Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103296.2 | NP_001096766.1 | 2 - 3 | 317 | 92 | |
GenBank mRNA | BC142333.1 | - | 2 - 3 | 318 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |