Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC49A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101287.1 | NP_001094757.1 | 3 - 4 | 801 | 97 | |
GenBank mRNA | BC148071.1 | - | 3 - 4 | 801 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |