Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102324.1 | NP_001095794.1 | 6 - 7 | 706 | 103 | |
GenBank mRNA | BC149374.1 | - | 6 - 7 | 706 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |