Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RACK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ132860.1 | - | 2 | 351 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Inventoried Amplicon spans exons and probe does not span exons |