Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144088.1 | NP_001137560.1 | 1 - 2 | 119 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |