Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC24A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144094.2 | NP_001137566.2 | 7 - 8 | 1539 | 101 | |
GenBank mRNA | EE340710.1 | - | 5 - 6 | 624 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |