Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205546.3 | NP_001192475.1 | 3 - 4 | 490 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |