Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2H2orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205626.2 | NP_001192555.2 | 6 - 7 | 532 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |