Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205688.1 | NP_001192617.1 | 2 - 3 | 447 | 80 | |
GenBank mRNA | EU616677.1 | - | 2 - 3 | 274 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |