Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205721.1 | NP_001192650.1 | 2 - 3 | 505 | 70 | |
GenBank mRNA | BC122594.1 | - | 2 - 3 | 371 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |