Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205862.1 | NP_001192791.1 | 1 - 2 | 879 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Bos taurus Probe spans exons |