Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206195.1 | NP_001193124.1 | 5 - 6 | 767 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |