Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192874.2 | NP_001179803.1 | 3 - 4 | 683 | 96 | |
GenBank mRNA | AJ868432.1 | - | 3 - 4 | 300 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |