Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166560.1 | NP_001160032.1 | 1 - 2 | 657 | 69 | |
GenBank mRNA | BC120369.1 | - | 1 - 2 | 464 | 69 | |
EE347275.1 | - | 1 - 2 | 657 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |