Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191183.1 | NP_001178112.1 | 4 - 5 | 793 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |