Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191195.1 | NP_001178124.1 | 1 - 2 | 826 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |