Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191198.2 | NP_001178127.2 | 5 - 6 | 473 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |