Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206822.1 | NP_001193751.1 | 18 - 19 | 3278 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |