Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206828.2 | NP_001193757.1 | 5 - 6 | 535 | 73 | |
GenBank mRNA | AJ676387.1 | - | 4 - 5 | 368 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |