Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206939.1 | NP_001193868.1 | 5 - 6 | 1593 | 70 | |
GenBank mRNA | EH177172.1 | - | 3 - 4 | 495 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |