Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244145.1 | NP_001231074.1 | 1121 | 66 | ||
GenBank mRNA | AF091342.1 | - | 1 - 2 | 729 | 66 | |
BC151624.1 | - | 1 - 2 | 1112 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |