Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191476.2 | NP_001178405.1 | 3 - 4 | 1191 | 69 | |
GenBank mRNA | EV635295.1 | - | 2 - 3 | 349 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |