Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191535.1 | NP_001178464.1 | 4 - 5 | 2495 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |