Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192149.1 | NP_001179078.1 | 2 - 3 | 136 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |