Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192287.3 | NP_001179216.2 | 1 - 2 | 479 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |