Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192363.2 | NP_001179292.1 | 5 - 6 | 1982 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |