Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192976.2 | NP_001179905.1 | 5 - 6 | 898 | 71 | |
GenBank mRNA | BC104525.2 | - | 5 - 6 | 834 | 71 | |
EH198200.1 | - | 2 - 3 | 232 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |