Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192980.3 | NP_001179909.3 | 1 - 2 | 413 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Inventoried Probe spans exons |