Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERTAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001354327.2 | NP_001341256.1 | 3 - 4 | 539 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Bos taurus Probe spans exons |