Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193259.1 | NP_001180188.1 | 5 - 6 | 569 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |