Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194961.1 | NP_001181890.1 | 1 | 720 | 80 | |
GenBank mRNA | GU936962.1 | - | 1 | 720 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |