Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046158.2 | NP_001039623.2 | 9 - 10 | 1269 | 71 | |
GenBank mRNA | BT025363.1 | - | 9 - 10 | 1197 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |