Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205307.3 | NP_001192236.2 | 16 - 17 | 3268 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |