Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205400.2 | NP_001192329.1 | 7 - 8 | 893 | 76 | |
GenBank mRNA | AF491875.1 | - | 4 - 5 | 510 | 76 | |
BC148117.1 | - | 7 - 8 | 894 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |