Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322663.4 | NP_001309674.1 | 6 - 7 | 1010 | 125 | |
NM_001322664.3 | NP_001309467.1 | 6 - 7 | 1010 | 125 | ||
NM_001322665.4 | NP_001309678.1 | 1 - 2 | 82 | 125 | ||
NM_001383675.1 | 6 - 7 | 1003 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |