Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029161.4 | NP_001024332.1 | 6 - 7 | 2100 | 71 | |
NM_001373600.3 | NP_001359587.1 | 6 - 7 | 2094 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |