Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375198.2 | NP_001361896.1 | 12 - 13 | 2095 | 69 | |
NM_001375199.1 | NP_001361830.1 | 12 - 13 | 2121 | 69 | ||
NM_001375200.2 | NP_001361897.1 | 11 - 12 | 1910 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Caenorhabditis elegans Probe spans exons |