Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sod-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078363.8 | NP_510764.1 | 1 - 2 | 105 | 70 | |
GenBank mRNA | X85790.1 | - | 65 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |